Please login to your online account to display your discounted pricing

Alfa Aesar™ O-tert-Butyl-L-threonine methyl ester hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material O-tert-Butyl-L-threonine methyl ester hydrochloride
CAS 71989-43-0
Formula Weight 225.71
MDL Number MFCD00077111
Beilstein 4716005
Synonym H-Thr(tBu)-Ome.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6352903

  SDS
alfa aesar™
H6352903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6352906

  SDS
alfa aesar™
H6352906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.71
Beilstein 4716005
Physical Form Solid
Quantity 1g
Chemical Name or Material O-tert-Butyl-L-threonine methyl ester hydrochloride
Assay 95%
Synonym H-Thr(tBu)-Ome.HCl
CAS 71989-43-0
Storage Note 1 2° to 8°C
MDL Number MFCD00077111
Safety and Handling

2° to 8°C