Please login to your online account to display your discounted pricing

Alfa Aesar™ O-tert-Butyl-L-serine tert-butyl ester hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material O-tert-Butyl-L-serine tert-butyl ester hydrochloride
CAS 51537-21-4
Formula Weight 253.77
MDL Number MFCD00034850
Beilstein 4886392
Synonym H-Ser(tBu)-OtBu.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6588714

  SDS
alfa aesar™
H6588714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6588706

  SDS
alfa aesar™
H6588706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 253.77
Melting Point 170° to 177°C
MDL Number MFCD00034850
Assay 97%
Chemical Name or Material O-tert-Butyl-L-serine tert-butyl ester hydrochloride
CAS 51537-21-4
Physical Form Crystalline Solid
Synonym H-Ser(tBu)-OtBu.HCl
Beilstein 4886392
Storage Note 1 2° to 8°C
Quantity 25g
Color White
Safety and Handling

2° to 8°C