missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-tert-Butyl-L-serine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6212603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
O-tert-Butyl-L-serine methyl ester hydrochloride | |
C8H18ClNO3 | |
2041979 | |
PCIABNBULSRKSU-UHFFFAOYNA-N | |
methyl 2-amino-3-(tert-butoxy)propanoate hydrochloride | |
16218549 | |
98% |
17114-97-5 | |
MFCD00077108 | |
h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl | |
Cl.COC(=O)C(N)COC(C)(C)C | |
211.69 | |
211.69 | |
1 g |
Chemical Identifiers
17114-97-5 | |
211.69 | |
PCIABNBULSRKSU-UHFFFAOYNA-N | |
16218549 |
C8H18ClNO3 | |
MFCD00077108 | |
h-ser tbu-ome.hcl, o-tert-butyl-l-serine methyl ester hydrochloride, h-ser tbu-ome hcl, s-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, methyl o-tert-butyl-l-serinate hydrochloride, l-serine, o-1,1-dimethylethyl-, methyl ester, hydrochloride, methyl 2s-2-amino-3-tert-butoxy propanoate hydrochloride, ksc491o3r, h-ser tbu-och3 hydrochloride, s-methyl 2-amino-3-tert-butoxy propanoate hcl | |
Cl.COC(=O)C(N)COC(C)(C)C |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only