missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-tert-Butyl-L-serine, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC434880050
Specifications
O-tert-Butyl-L-serine | |
− 15.50 | |
White | |
5g | |
96% min. (HPLC) | |
2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, 2s-2-amino-3-tert-butoxy propanoic acid, h-ser but-oh, h-ser tbu-oh, l-serine, o-1,1-dimethylethyl, o-t-butyl-l-serine, o-tert-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, ser tbu-oh | |
CC(C)(C)OCC(C(=O)O)N | |
161.2 | |
161.2 | |
97% |
Authentic | |
10% Max. (K.F.) | |
225.2°C to 228.0°C | |
18822-58-7 | |
C7H15NO3 | |
DDCPKNYKNWXULB-YFKPBYRVSA-N | |
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
7004940 | |
Crystalline Powder |
Chemical Identifiers
18822-58-7 | |
161.2 | |
2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, 2s-2-amino-3-tert-butoxy propanoic acid, h-ser but-oh, h-ser tbu-oh, l-serine, o-1,1-dimethylethyl, o-t-butyl-l-serine, o-tert-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, ser tbu-oh | |
(2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
C7H15NO3 | |
DDCPKNYKNWXULB-YFKPBYRVSA-N | |
7004940 | |
CC(C)(C)OCC(C(=O)O)N |