missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-tert-Butyl-D-serine methyl ester hydrochloride, 98%

Manufacturer:  Alfa Aesar™ H6387406

 View more versions of this product

Catalog No. AAH6387406



Specifications

Specifications

O-tert-Butyl-D-serine methyl ester hydrochloride
C8H18ClNO3
2041979
PCIABNBULSRKSU-FYZOBXCZSA-N
methyl (2R)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
13571432
Powder
White
78537-14-1
MFCD00077108
h-d-ser tbu-ome.hcl, h-d-ser tbu-ome hydrochloride, o-t-butyl-d-serine methyl ester hydrochloride, ser tbu-ome.hcl, r-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, h-d-ser tbu-ome hcl, o-tert-butyl-d-serine methyl ester hydrochloride, methyl o-tert-butyl-d-serinate-hydrogen chloride 1/1, methyl 2r-2-amino-3-tert-butoxy propanoate hydrochloride
CC(C)(C)OCC(C(=O)OC)N.Cl
211.686
211.69
98%
5g

Chemical Identifiers

78537-14-1
211.686
PCIABNBULSRKSU-FYZOBXCZSA-N
13571432
CC(C)(C)OCC(C(=O)OC)N.Cl
C8H18ClNO3
MFCD00077108
h-d-ser tbu-ome.hcl, h-d-ser tbu-ome hydrochloride, o-t-butyl-d-serine methyl ester hydrochloride, ser tbu-ome.hcl, r-methyl 2-amino-3-tert-butoxy propanoate hydrochloride, h-d-ser tbu-ome hcl, o-tert-butyl-d-serine methyl ester hydrochloride, methyl o-tert-butyl-d-serinate-hydrogen chloride 1/1, methyl 2r-2-amino-3-tert-butoxy propanoate hydrochloride
methyl (2R)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoate;hydrochloride
SDS Documents