missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Terphenyl, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1968014

 View more versions of this product

Catalog No. AAA1968014



Description

Description

Applications
Terphenyl mixtures are used industrially as heat storage and transfer agents, as textile dye carriers, and as intermediates in the production of nonspreading lubricants. Contemporary use of o-Terphenyl is found in solar-heating systems. It is used as a plasticizer for polystyrene for thermoplastic recording.

Solubility
Insoluble in water. Solubility in toluene is almost transparent. Sparingly soluble in lower alcohols and glycols; very soluble in common aromatic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

o-Terphenyl
25g
0.98
MFCD00003055
1908446
OIAQMFOKAXHPNH-UHFFFAOYSA-N
1,2-diphenylbenzene
6766
98%
1.100
84-15-1
C18H14
UN3077
o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655
C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3
230.31
230.31

Chemical Identifiers

84-15-1
230.31
OIAQMFOKAXHPNH-UHFFFAOYSA-N
6766
C1=CC=C(C=C1)C2=CC=CC=C2C3=CC=CC=C3
C18H14
MFCD00003055
o-terphenyl, 1,1':2',1-terphenyl, terphenyl, chlorinated, 1,1'-biphenyl, 2-phenyl, caswell no. 672b, o-diphenylbenzene, polychlorinated terphenyl, polychlorinated terphenyls, unii-w5675r7kvw, ccris 1655
1,2-diphenylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : WZ6472000

TSCA : TSCA

SDS Documents