Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Terphenyl, 98%

$56.50 - $145.54

Chemical Identifiers

Chemical Name or Material o-Terphenyl
CAS 84-15-1
Molecular Formula C18H14
Formula Weight 230.31
MDL Number MFCD00003055
UN Number UN3077
Beilstein 1908446
Synonym 1,2-Diphenylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1968022

  SDS
alfa aesar™
A1968022
100g Each for $145.54

AAA1968014

  SDS
alfa aesar
A1968014
25g Each for $56.50
Description & Specifications

Specifications

Formula Weight 230.31
Assay Percent Range 0.98
Melting Point 56° to 58°C
Synonym 1,2-Diphenylbenzene
Boiling Point 332°C
TSCA TSCA
Health Hazard 1 Warning
MDL Number MFCD00003055
UN Number UN3077
CAS 84-15-1
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number WZ6472000
Chemical Name or Material o-Terphenyl
Quantity 100g
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Molecular Formula C18H14
Flash Point 163°C (325°F)
Beilstein 1908446
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Terphenyl mixtures are used industrially as heat storage and transfer agents, as textile dye carriers, and as intermediates in the production of nonspreading lubricants. Contemporary use of o-Terphenyl is found in solar-heating systems. It is used as a plasticizer for polystyrene for thermoplastic recording.

Solubility
Insoluble in water. Solubility in toluene is almost transparent. Sparingly soluble in lower alcohols and glycols; very soluble in common aromatic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.