Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Succinimidyl-1,3-dimethylpropyleneuronium hexafluorophosphate, 98%

$192.59 - $192.59

Chemical Identifiers

Chemical Name or Material O-Succinimidyl-1,3-dimethylpropyleneuronium hexafluorophosphate
CAS 443305-33-7
Molecular Formula C10H16F6N3O3P
Formula Weight 419.26g/mol
MDL Number MFCD08274619
Synonym HDP-OSu
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2641806

 
alfa aesar™
H26418-06
5g Each for $192.59
Description & Specifications

Specifications

Melting Point 172° to 175°C
Quantity 5g
Chemical Name or Material O-Succinimidyl-1,3-dimethylpropyleneuronium hexafluorophosphate
CAS 443305-33-7
Assay 98%
Molecular Formula C10H16F6N3O3P
Formula Weight 419.26g/mol
MDL Number MFCD08274619
Synonym HDP-OSu
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C