Learn More
O,S-Diethyl methylphosphonothioate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04446502
Description
O,S-Diethyl methylphosphonothioate is used as an inhibitor of cholinesterase. It is also used in pharmaceuticals, intermediates and fine chemicals. It also serves as phosphorylating and phosphitylating agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
O,S-Diethyl methylphosphonothioate | |
1.094 g/mL | |
C5H13O2PS | |
MFCD01705977 | |
UN3278 | |
Immiscible with water. | |
CCOP(=O)(C)SCC | |
168.19 | |
168.19 | |
Liquid |
2511-10-6 | |
95°C to 97°C (10 mmHg) | |
1.474 | |
0.5 g | |
o,s-diethyl methylphosphonothioate, o,s-diethyl methylthiophosphonate, methylphosphonothioic acid o,s-diethyl ester, osdmp, phosphonothioic acid, methyl-, o,s-diethyl ester, phosphonothioic acid, p-methyl-, o,s-diethyl ester, o-ethyl s-ethyl methylphosphonothioate, ethyl ethylsulfanyl methyl phosphinate, acmc-20ak0l, ethoxy ethylthio methyl phosphine oxide | |
AIMOGGQLCCAPQJ-UHFFFAOYNA-N | |
1-[ethylsulfanyl(methyl)phosphoryl]oxyethane | |
520177 | |
97% |
Chemical Identifiers
2511-10-6 | |
MFCD01705977 |
Safety and Handling
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
RTECSNumber : TB1085000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only