missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-Phosphorylethanolamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367420250

 View more versions of this product

Catalog No. AC367420250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

O-Phosphorylethanolamine
1071-23-4
97.5 % min. (Titration with HClO4)
C2H8NO4P
MFCD00008178
04,IV,1415
Solubility in water: soluble.
C(COP(=O)(O)O)N
141.06
CHEBI:17553
98%
98%
241.0°C to 243.0°C
Glass Bottle
H2NCH2CH2OP(O)(OH)2
25 g
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
SUHOOTKUPISOBE-UHFFFAOYSA-N
2-aminoethyl dihydrogen phosphate
1015
141.06

Chemical Identifiers

1071-23-4
141.06
SUHOOTKUPISOBE-UHFFFAOYSA-N
1015
2-aminoethyl dihydrogen phosphate
C2H8NO4P
MFCD00008178
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
CHEBI:17553
C(COP(=O)(O)O)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS Signal Word: Danger

EINECSNumber : 213-988-5

Recommended Storage : Refrigerator +4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only