Please login to your online account to display your discounted pricing

o-Phosphoric Acid (NF/FCC), Fisher Chemical

$299.92 - $912.46

Chemical Identifiers

Chemical Name or Material o-Phosphoric Acid
CAS 7664-38-2, 7732-18-5
CAS 7732-18-5
Purity Grade FCC/NF
Assay 85.0 to 88.0% (NF)
Molecular Formula H3O4P
Formula Weight 98
MDL Number MFCD00011340
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A365-1

  Accessories SDS
fisher scientific company
BPA3651
Glass Bottle 1L Each for $299.92
Case of 6 Each for $1,799.52

A365-4

  Accessories SDS
fisher scientific company
BPA3654
Glass Bottle 4L Each for $912.46
Case of 4 Each for $3,649.84
Description & Specifications

Specifications

Purity Grade FCC/NF
Formula Weight 98
Melting Point 21°C
Boiling Point 158°C
Quantity 1L
CAS 7664-38-2, 7732-18-5
UN Number 1805
DOT Information DOT Class 8, : Corrosive
Assay 85.0 to 88.0% (FCC)
Physical Form Liquid
Color Clear Viscous
pH 1
Vapor Pressure 2 hPa at 20°C
Density 1.680g/cm³
Alpha Vector Phosphoric Acid
Identification Pass Test (FCC)
Arsenic (As) 3ppm max. (FCC)
CAS 7732-18-5
Heavy Metals (as Pb) 0.001% max. (NF)
Fluorides 10mg/kg max. (FCC)
Health Hazard 3 Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:0
Nitrate Pass Test (NF)
MDL Number MFCD00011340
Cleanup Symbol Acid
Alkali Phosphates Pass Test (NF)
Sulfate Pass Test (NF)
Lead (Pb) 3mg/kg max. (FCC)
Packaging Glass Bottle
Phosphorous and Hypophosphorous Acid Pass Test (NF)
Residual Solvents Meets Requirements (NF)
Cadmium (Cd) 3mg/kg max. (FCC)
Health Hazard 2 DANGER!
ChemAlert Storage Symbol White
Molecular Formula H3O4P
Chemical Name or Material o-Phosphoric Acid

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:0

DANGER!