missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Phenylenediamine, Fisher BioReagents

Free Base

$130.46 - $205.37

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym 1,2-benzenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP253750 SDS View Documents Fisher BioReagents
BP253750
50g Amber Glass Each for $130.46
Case of 8 Each for $1,043.68
 
BP2537250 SDS View Documents Fisher BioReagents
BP2537250
250g Amber Glass Each for $205.37
Case of 4 Each for $821.48
 
Description

Description

  • A chromogenic peroxidase substrate useful for ELISA and other enzyme immunoassays
  • Commonly used as a substrate for horseradish peroxidase (HRP) in ELISAs
  • When oxidized, OPD produces a color change that is detectable by spectrophotometry
  • Color intensity will increase over time as OPD is oxidzed by peroxidase
Specifications

Chemical Identifiers

95-54-5
108.144
1,2-benzenediamine
CHEBI:34043
C1=CC=C(C(=C1)N)N
C6H8N2
GEYOCULIXLDCMW-UHFFFAOYSA-N
7243
benzene-1,2-diamine

Specifications

Amber Glass
Cream
50g
C6H8N2
15, 7396
GEYOCULIXLDCMW-UHFFFAOYSA-N
benzene-1,2-diamine
7243
108.1
o-Phenylenediamine
256°C
100°C
95-54-5
C6H4(NH2)2
1,2-benzenediamine
C1=CC=C(C(=C1)N)N
108.144
CHEBI:34043
Solid
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

Recommended Storage : RT