Learn More
o-Phenylenediamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130550050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
o-Phenylenediamine | |
97.5 | |
100.0°C to 103.0°C | |
256.0°C to 258.0°C | |
Authentic | |
Plastic drum | |
C6H4(NH2)2 | |
5 kg | |
01,834; 08,393 | |
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine | |
GEYOCULIXLDCMW-UHFFFAOYSA-N | |
benzene-1,2-diamine | |
7243 | |
108.14 | |
Crystalline Powder |
95-54-5 | |
100.0 | |
White to Beige | |
136°C | |
97.5% min. (GC) | |
C6H8N2 | |
MFCD00007721 | |
13, IV, 38 | |
15, 7396 | |
Solubility in water: 54g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform | |
C1=CC=C(C(=C1)N)N | |
108.14 | |
CHEBI:34043 | |
98% |
Chemical Identifiers
95-54-5 | |
108.14 | |
GEYOCULIXLDCMW-UHFFFAOYSA-N | |
7243 | |
benzene-1,2-diamine |
C6H8N2 | |
MFCD00007721 | |
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine | |
CHEBI:34043 | |
C1=CC=C(C(=C1)N)N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a posit
GHS Signal Word: Danger
EINECSNumber : 202-430-6
Recommended Storage : This product tends to darken during storage
RUO – Research Use Only