Please login to your online account to display your discounted pricing

o-Phenetidine, 99%, ACROS Organics™

$24.33 - $249.17

Chemical Identifiers

Chemical Name or Material o-Phenetidine
Name Note 99%
CAS 94-70-2
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H11NO
Linear Formula C2H5OC6H4NH2
Formula Weight 137.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC233480050

  SDS
thermo fisher scientific
233480050
Glass bottle 5mL Each for $24.33

AC233485000

  SDS
thermo fisher scientific
233485000
Glass bottle 500mL Each for $249.17
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 137.18
Quantity 5mL
Color Deep red
EINECS Number 202-356-4
TSCA TSCA
Density 1.05g/mL
RTECS Number SI6465300
Merck Index 15, 7341
CAS Max % 100.0
Chemical Name or Material o-Phenetidine
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H11NO
Refractive Index 1.5540 to 1.5560 (20°C, 589nm)
Linear Formula C2H5OC6H4NH2
CAS 94-70-2
Synonym 2-Ethoxybenzenamine
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99%
Assay Percent Range 99%
MDL Number MFCD00007689
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.