Please login to your online account to display your discounted pricing

o-Phenanthroline Solution, Certified, 0.25% (w/v), LabChem™

$81.14 - $81.14

Chemical Identifiers

Chemical Name or Material o-Phenanthroline Solution
CAS 7732-18-5
CAS Max % 99.75
CAS 5144-89-8
CAS Max % 0.25
Purity Grade Certified Grade
Linear Formula C12H8N2·H2O
Formula Weight 198.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC181601

 
lab chem
LC181601
500mL Each for $81.14
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 198.22
Specific Gravity 1g/mL
Storage Information Room Temperature
CAS Max % 99.75
Suitability Passes Test
Physical Form Liquid
Density 1g/mL
Color Clear to almost colorless
CAS Max % 0.25
Health Hazard 2 GHS H Statement
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Assay 0.25% (w/v)
CAS 5144-89-8
Solubility Soluble in water
Decomposition Information Carbon monoxide; Carbon dioxide
Chemical Name or Material o-Phenanthroline Solution
Identification Passes Test
Quantity 500mL
Linear Formula C12H8N2·H2O
CAS 7732-18-5
Packaging Poly Bottle
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life with long lasting effects.