Please login to your online account to display your discounted pricing

o-Phenanthroline, 0.3% (w/v), LabChem™

$168.20 - $168.20

Chemical Identifiers

Chemical Name or Material o-Phenanthroline
CAS 7732-18-5
CAS Max % 99.6
CAS 5144-89-8
CAS Max % 0.3
CAS 7647-01-0
CAS Max % 0.1
Linear Formula C12H8N2·H2O
Formula Weight 198.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC181552

 
lab chem
LC181552
Poly Bottle 1L Each for $168.20
Description & Specifications

Specifications

Formula Weight 198.22
Chemical Name or Material o-Phenanthroline
CAS Max % 0.1
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Assay 0.3% (w/v)
Linear Formula C12H8N2·H2O
CAS 7732-18-5
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Density 1g/mL
CAS Max % 99.6
Health Hazard 2 GHS H Statement
Harmful to aquatic life with long lasting effects.
Color Clear to almost colorless
CAS 5144-89-8
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.3
CAS 7647-01-0
Quantity 1L
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life with long lasting effects.