Please login to your online account to display your discounted pricing

o-Phenanthroline , 0.1% Aqueous, for Iron, Certified, LabChem™

$79.03 - $79.03

Chemical Identifiers

Chemical Name or Material o-Phenanthroline
CAS 7732-18-5
CAS Max % 99.8
CAS 5144-89-8
CAS Max % 0.1
CAS 7647-01-0
CAS Max % 0.1
Name Note 0.1% Aqueous, for Iron
Purity Grade Certified Grade
Linear Formula C12H8N2·H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC181501

 
lab chem
LC181501
500mL Each for $79.03
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 198.22
Chemical Name or Material o-Phenanthroline
CAS Max % 0.1
Decomposition Information Carbon monoxide; Carbon dioxide
Color Clear to almost colorless
Quantity 500mL
Name Note 0.1% Aqueous, for Iron
Physical Form Liquid
Linear Formula C12H8N2·H2O
Density 1g/mL
CAS 7732-18-5
CAS Max % 99.8
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 5144-89-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 0.1
Solubility Soluble in water
CAS 7647-01-0
Specific Gravity 1g/mL
Suitability Passes Test
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.