Please login to your online account to display your discounted pricing

o-Phenanthroline, 0.1% Alcoholic, Certified, LabChem™

$53.51 - $53.51

Chemical Identifiers

Chemical Name or Material o-Phenanthroline
CAS 64-17-5
CAS Max % 89.91
CAS 67-63-0
CAS Max % 4.995
CAS 67-56-1
CAS Max % 4.995
CAS 5144-89-8
CAS Max % 0.1
Name Note 0.1% Alcoholic
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC181701

 
lab chem
LC181701
Poly Bottle 500mL Each for $53.51
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 198.22
Chemical Name or Material o-Phenanthroline
CAS Max % 4.995
CAS 5144-89-8
CAS Max % 0.1
Identification Passes Test
Name Note 0.1% Alcoholic
Packaging Poly Bottle
Physical Form Liquid
Linear Formula C12H8N2·H2O
CAS 64-17-5
UN Number UN1170
Solubility Soluble in water
Storage Information Room Temperature
Suitability Passes Test
Health Hazard 1 Danger
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
CAS Max % 89.91
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral).
Color Clear to almost colorless
CAS 67-63-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 4.995
CAS 67-56-1
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral).