missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ (-)-O,O'-Dipivaloyl-L-tartaric acid, 98+%
Manufacturer: Alfa Aesar™ H5603203
Specifications
(-)-O,O'-Dipivaloyl-L-tartaric acid | |
C14H22O8 | |
2008171 | |
UFHJEZDFEHUYCR-YUMQZZPRSA-N | |
(2S,3S)-2,3-bis(2,2-dimethylpropanoyloxy)butanedioic acid | |
16212279 | |
≥98% | |
1g | |
Hygroscopic |
65259-81-6 | |
MFCD00066535 | |
+-dipivaloyl-d-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, +-o,o'-di-pivaloyl-d-tartaric acid, --bis 2,2-dimethylpropionyl-l-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, ?-o,o'-di-pivaloyl-l-tartaric acid, d-+-dipivaloyl tartaric acid, dipivaloyl-l-tartaric acid, pubchem8173 | |
CC(C)(C)C(=O)OC(C(C(=O)O)OC(=O)C(C)(C)C)C(=O)O | |
318.322 | |
318.32 | |
127°C to 132°C | |
−24° (c=1.7 in Dioxane) |
Chemical Identifiers
65259-81-6 | |
318.322 | |
UFHJEZDFEHUYCR-YUMQZZPRSA-N | |
16212279 | |
CC(C)(C)C(=O)OC(C(C(=O)O)OC(=O)C(C)(C)C)C(=O)O |
C14H22O8 | |
MFCD00066535 | |
+-dipivaloyl-d-tartaric acid, +-dipivaloyl-d-tartaricacid, +-dpta, +-o,o'-di-pivaloyl-d-tartaric acid, --bis 2,2-dimethylpropionyl-l-tartaric acid, 2s,3s-2,3-bis pivaloyloxy succinic acid, ?-o,o'-di-pivaloyl-l-tartaric acid, d-+-dipivaloyl tartaric acid, dipivaloyl-l-tartaric acid, pubchem8173 | |
(2S,3S)-2,3-bis(2,2-dimethylpropanoyloxy)butanedioic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen