missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O,O'-Bis(trimethylsilyl)-5-fluorouracil, 97%

$119.36 - $373.19

Chemical Identifiers

CAS 17242-85-2
Molecular Formula C10H19FN2O2Si2
Molecular Weight (g/mol) 274.442
MDL Number MFCD00042441
InChI Key MPVQFVDWJDULDL-UHFFFAOYSA-N
Synonym 2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
PubChem CID 140203
IUPAC Name (5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane
SMILES C[Si](C)(C)OC1=NC(=NC=C1F)O[Si](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1640706 View Documents Alfa Aesar™
L1640706
5g Each for $119.36
 
AAL1640714 View Documents Alfa Aesar™
L1640714
25g Each for $373.19
 
Specifications

Chemical Identifiers

17242-85-2
274.442
MPVQFVDWJDULDL-UHFFFAOYSA-N
140203
C[Si](C)(C)OC1=NC(=NC=C1F)O[Si](C)(C)C
C10H19FN2O2Si2
MFCD00042441
2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
(5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane

Specifications

32°C (89°F)
5g
C10H19FN2O2Si2
UN1992
2,4-bis trimethylsilyl-5-fluorouracil, o,o'-bis trimethylsilyl-5-fluorouracil, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 5-fluoro-2,4-bis trimethylsilyl oxy pyrimidine, 2,4-bis trimethylsilyloxy-5-fluoropyrimidine, 5-fluorouracil, tms, uracil, 5-fluoro, tms, 2,4-bis trimethylsilyl-5-fluorouraci, 5-fluoro-2,4-bis trimethylsilyloxy pyrimidine, 2,4-di-trimethylsilyloxy-5-fluoropyrimidine
C[Si](C)(C)OC1=NC(=NC=C1F)O[Si](C)(C)C
274.442
274.44
O,O'-Bis(trimethylsilyl)-5-fluorouracil
114°C (14mmHg)
17242-85-2
MFCD00042441
885268
MPVQFVDWJDULDL-UHFFFAOYSA-N
(5-fluoro-2-trimethylsilyloxypyrimidin-4-yl)oxy-trimethylsilane
140203
97%
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H341-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.