Learn More
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 303190050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Coupling reagentSpecifications
O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate | |
96.0 | |
34.47 to 37.34% C, 13.40 to 14.52% N | |
97% | |
C9H16N3O3·BF4 | |
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781 | |
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O | |
301.05 | |
301.05 |
105832-38-0 | |
100.0 | |
Authentic | |
Glass bottle | |
5g | |
YEBLHMRPZHNTEK-UHFFFAOYSA-N | |
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate | |
9857522 | |
97% |
Chemical Identifiers
105832-38-0 | |
301.05 | |
tstu, o-n-succinimidyl-1,1,3,3-tetramethyluronium tetrafluoroborate, 2-2,5-dioxopyrrolidin-1-yl-1,1,3,3-tetramethylisouronium tetrafluoroborate, 2-succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, n,n,n',n'-tetramethyl-o-n-succinimidyl uronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyluronium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl-uronium tetrafluoroborate, dimethylamino 2,5-dioxopyrrolidin-1-yl oxy methylidene dimethylazanium tetrafluoroborate, o-n-succinimidyl-n,n,n',n'-tetramethyl uronium bf4, pubchem12781 | |
[dimethylamino-(2,5-dioxopyrrolidin-1-yl)oxymethylidene]-dimethylazanium;tetrafluoroborate |
C9H16N3O3·BF4 | |
YEBLHMRPZHNTEK-UHFFFAOYSA-N | |
9857522 | |
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)CCC1=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON
GHS Signal Word: Warning
RUO â Research Use Only