missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Methylisourea hemisulfate, 94%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 233032500
Specifications
O-Methylisourea hemisulfate | |
147°C to 153°C | |
94% | |
92.0 | |
94% | |
0·5H2SO4 | |
0-methylisourea hemisulfate, 2-methylisouronium sulfate, bis 2-methylisouronium sulphate, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, o-methylisourea hemisulfate, o-methylisourea hemisulfate salt, o-methylisourea hemisulphate, o-methylpseudourea sulfate, omi™ | |
QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
methyl carbamimidate;sulfuric acid | |
11357018 | |
94% |
Plastic bottle | |
250g | |
52328-05-9 | |
100.0 | |
0·5 H2SO4 | |
MFCD00040594 | |
Solubility in water: 1000g/L water (20°C) | |
COC(=N)N.COC(=N)N.OS(=O)(=O)O | |
123.12 | |
123.12 |
Chemical Identifiers
52328-05-9 | |
123.12 | |
QSCPQKVWSNUJLJ-UHFFFAOYSA-N | |
11357018 | |
COC(=N)N.COC(=N)N.OS(=O)(=O)O |
0·5 H2SO4 | |
MFCD00040594 | |
0-methylisourea hemisulfate, 2-methylisouronium sulfate, bis 2-methylisouronium sulphate, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, o-methylisourea hemisulfate, o-methylisourea hemisulfate salt, o-methylisourea hemisulphate, o-methylpseudourea sulfate, omi™ | |
methyl carbamimidate;sulfuric acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 257-851-8