missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-Methylisourea hemisulfate, 94%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 233030250

 View more versions of this product

Catalog No. AC233030250



Specifications

Specifications

O-Methylisourea hemisulfate
147°C to 153°C
94%
92.0
94%
0·5H2SO4
0-methylisourea hemisulfate
QSCPQKVWSNUJLJ-UHFFFAOYSA-N
methyl carbamimidate;sulfuric acid
11357018
94%
Plastic bottle
25g
52328-05-9
100.0
0·5 H2SO4
MFCD00040594
Solubility in water: 1000g/L water (20°C)
COC(=N)N.COC(=N)N.OS(=O)(=O)O
123.12
123.12

Chemical Identifiers

52328-05-9
123.12
QSCPQKVWSNUJLJ-UHFFFAOYSA-N
11357018
COC(=N)N.COC(=N)N.OS(=O)(=O)O
0·5 H2SO4
MFCD00040594
0-methylisourea hemisulfate
methyl carbamimidate;sulfuric acid
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 257-851-8

SDS Documents