Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Methyl-D-serine hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material O-Methyl-D-serine hydrochloride
CAS 86118-10-7
Formula Weight 155.58
MDL Number MFCD11099371
Beilstein 56965747
Synonym H-Ser(Me)-OH.HCl; (R)-2-Amino-3-methoxypropionic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6588403

 
alfa aesar™
H6588403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65884MD

 
alfa aesar™
H65884MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6588406

 
alfa aesar™
H6588406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 155.58
CAS 86118-10-7
Physical Form Solid
Assay 95%
MDL Number MFCD11099371
Synonym H-Ser(Me)-OH.HCl; (R)-2-Amino-3-methoxypropionic acid hydrochloride
Beilstein 56965747
Quantity 1g
Sensitivity Hygroscopic
Chemical Name or Material O-Methyl-D-serine hydrochloride