Please login to your online account to display your discounted pricing

o-Fluorotoluene ≥99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  119660100

 View more versions of this product

Catalog No. AC119660100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 110.13
Quantity 10g
Color Clear colorless to slightly yellow
Merck Index 15, 4212
Beilstein 05, 290
TSCA TSCA
RTECS Number XT2579000
EINECS Number 202-428-5
Density 1g/mL
CAS Max % 100.0
Linear Formula CH3C6H4F
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
CAS Min % 98.5
Packaging Glass bottle
Refractive Index 1.4720 to 1.4750 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Chemical Name or Material o-Fluorotoluene
MDL Number MFCD00000322
CAS 95-52-3
Molecular Formula C7H7F
Assay Percent Range 99%

Specifications

Chemical Name or Material o-Fluorotoluene
Name Note 99+%
CAS 95-52-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H7F
Linear Formula CH3C6H4F
Formula Weight 110.13
MDL Number MFCD00000322
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents