missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-Ethyl-N,N′-diisopropylisourea, TCI America™

Manufacturer:  TCI America E105825ML

 View more versions of this product

Catalog No. E105825ML



Specifications

Specifications

O-Ethyl-N,N′-diisopropylisourea
Colorless
60683-30-9
Ethyl N,N′-Diisopropylcarbamimidate, N,N′-Diisopropylcarbamimidic Acid Ethyl Ester
CCOC(=NC(C)C)NC(C)C
172.272
172.27
≥98.0% (GC,T)
65°C
25mL
C9H20N2O
AUCYHQMYJIEKBO-UHFFFAOYSA-N
ethyl N,N'-di(propan-2-yl)carbamimidate
20470583
Clear Liquid at 20°C

Chemical Identifiers

60683-30-9
172.272
Ethyl N,N′-Diisopropylcarbamimidate, N,N′-Diisopropylcarbamimidic Acid Ethyl Ester
ethyl N,N'-di(propan-2-yl)carbamimidate
C9H20N2O
AUCYHQMYJIEKBO-UHFFFAOYSA-N
20470583
CCOC(=NC(C)C)NC(C)C
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents