missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ H5419406

 View more versions of this product

Catalog No. AAH5419406



Specifications

Specifications

O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium tetrafluoroborate
216°C to 219°C
125700-73-4
MFCD00167753
DMTWWFCCBWWXSA-NIQYDHDSSA-N
365.136
365.13
Light and moisture sensitive
5g
C14H20BF4N3O3
2-endo-5-norbornene-2,3-dicarboxymido-1,1,3,3-tetramethyluronium tetrafluoroborate
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)C2C3CC(C2C1=O)C=C3
91634054
≥98%

Chemical Identifiers

125700-73-4
365.136
DMTWWFCCBWWXSA-NIQYDHDSSA-N
91634054
C14H20BF4N3O3
MFCD00167753
2-endo-5-norbornene-2,3-dicarboxymido-1,1,3,3-tetramethyluronium tetrafluoroborate
[B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C(=O)C2C3CC(C2C1=O)C=C3
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents