Please login to your online account to display your discounted pricing

Alfa Aesar™ O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%

$137.61 - $137.61

Chemical Identifiers

Chemical Name or Material O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium hexafluorophosphate
CAS 208462-94-6
Molecular Formula C14H20F6N3O3P
Formula Weight 423.30g/mol
MDL Number MFCD08274617
Synonym HNTU; O-(Bicyclo[2.2.1]hept-5-ene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium hexafluorophosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2656806

 
alfa aesar™
H26568-06
5g Each for $137.61
Description & Specifications

Specifications

Melting Point 214° to 216°C
Quantity 5g
Chemical Name or Material O-(endo-5-Norbornene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium hexafluorophosphate
CAS 208462-94-6
Assay 98%
Molecular Formula C14H20F6N3O3P
Formula Weight 423.30g/mol
MDL Number MFCD08274617
Synonym HNTU; O-(Bicyclo[2.2.1]hept-5-ene-2,3-dicarboximido)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C