missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ o-Cresolphthalein complexone tetrasodium salt

$38.18 - $366.35

Chemical Identifiers

CAS 62698-54-8
Molecular Formula C32H28N2Na4O12
Formula Weight 724.54
MDL Number MFCD00066371
Synonym o-Cresolphthalexon, water soluble; Phthalein complexone sodium salt
Chemical Name or Material o-Cresolphthalein complexone tetrasodium salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1794103 SDS Alfa Aesar™
A1794103
1g Each for $38.18
AAA1794114 SDS Alfa Aesar™
A1794114
25g Each for $366.35
AAA1794106 SDS Alfa Aesar™
A1794106
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

62698-54-8
724.54
o-Cresolphthalexon, water soluble; Phthalein complexone sodium salt
C32H28N2Na4O12
MFCD00066371
o-Cresolphthalein complexone tetrasodium salt

Specifications

1g
C32H28N2Na4O12
MFCD00066371
o-Cresolphthalein complexone tetrasodium salt
62698-54-8
724.54
o-Cresolphthalexon, water soluble; Phthalein complexone sodium salt

Safety and Handling

TSCA : TSCA