missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Cresol, 99%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC405738000

 View more versions of this product

Catalog No. AC405738000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

o-Cresol
95-48-7,108-95-2
100.0
Authentic
Liquid or Low Melting Solid
99%
CH3C6H4OH
MFCD00002226
0.2% max.
Colorless to amber
800mL
98.5
1.0400g/mL
AcroSeal™ Glass Bottle
≥98.5%
C7H8O
108.14
Darkens on storage and exposure to light and air
2-Hydroxytoluene

Chemical Identifiers

95-48-7,108-95-2
100.0
C7H8O
108.14
2-Hydroxytoluene
≥98.5%
98.5
99%
CH3C6H4OH
MFCD00002226
o-Cresol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger