missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Cresol, 99%, ACROS Organics™

Manufacturer:  Acros Organics 110555000

 View more versions of this product

Catalog No. AC110555000



Specifications

Specifications

o-Cresol
81°C
Glass bottle
191.0°C
30.0°C to 32.0°C
95-48-7,108-95-2
100.0
C7H8O
MFCD00002226
QWVGKYWNOKOFNN-UHFFFAOYSA-N
2-methylphenol
335
108.14
99%
1.0400g/mL
Authentic
0.2% max. (K.F.)
Amber to Colorless
500g
98.5
99%
CH3C6H4OH
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
CC1=CC=CC=C1O
108.14
CHEBI:28054
Liquid or Low Melting Solid

Chemical Identifiers

95-48-7
108.14
QWVGKYWNOKOFNN-UHFFFAOYSA-N
335
2-methylphenol
C7H8O
MFCD00002226
o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
CHEBI:28054
CC1=CC=CC=C1O
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Recommended Storage : Darkens on storage and exposure to light and air

SDS Documents