missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-bromophenoxyacetic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 1879-56-7
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H7BrO3
Linear Formula BrC6H4OCH2CO2H
Formula Weight 231.05
MDL Number MFCD00234407
Chemical Name or Material 2-Bromophenoxyacetic acid
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC379210250 SDS Thermo Fisher Scientific
AC379210250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1879-56-7
100.0
C8H7BrO3
231.05
2-Bromophenoxyacetic acid
97.5
98%
BrC6H4OCH2CO2H
MFCD00234407
≥97.5%

Specifications

1879-56-7
100.0
Glass bottle
25g
Crystalline Powder
C8H7BrO3
231.05
2-Bromophenoxyacetic acid
97.5
Authentic
141° to 145°C
White to light pink
98%
BrC6H4OCH2CO2H
MFCD00234407
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning