Learn More
O-Benzylhydroxylamine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1360114
Description
O-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-benzylhydroxylamine used in the synthesis of alfa- hydroxybenzylamines from alfa-hydroxyketones. It is also involved in the preparation of hydroxylamines and hydroxyamates.
Solubility
Soluble in dimethylsulfoxide and methanol.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
O-Benzylhydroxylamine | |
1.058 | |
C7H9NO | |
C6H5CH2ONH2 | |
25 g | |
1906691 | |
Soluble in dimethylsulfoxide and methanol. | |
C1=CC=C(C=C1)CON | |
123.155 | |
123.15 |
622-33-3 | |
118°C to 119°C (30 mmHg) | |
1.541 | |
MFCD00221709 | |
UN2735 | |
benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine | |
XYEOALKITRFCJJ-UHFFFAOYSA-N | |
O-benzylhydroxylamine | |
102313 | |
96% |
Chemical Identifiers
622-33-3 | |
123.155 | |
XYEOALKITRFCJJ-UHFFFAOYSA-N | |
102313 | |
C1=CC=C(C=C1)CON |
C7H9NO | |
MFCD00221709 | |
benzyloxyamine, hydroxylamine, o-phenylmethyl, hydroxylamine, o-benzyl, o-benzyl-hydroxylamine, benzyl-o-hydroxylamine, oxybenzylamine, o-phenylmethyl hydroxylamine, o-benzyloxylamine, phenylmethoxyamine, benzyloxy amine | |
O-benzylhydroxylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only