missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-N-benzyloxycarbonyl-L-threonine, 95%

$34.55 - $619.93

Chemical Identifiers

CAS 69863-36-1
Molecular Formula C19H21NO5
Molecular Weight (g/mol) 343.379
MDL Number MFCD00077042
InChI Key FVKJXAYBJIAXAU-PBHICJAKSA-N
Synonym z-thr bzl-oh, n-z-o-benzyl-l-threonine, n-cbz-o-benzyl-l-threonine, cbz-thr bzl-oh, o-benzyl-n-benzyloxycarbonyl-l-threonine, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, ambotzzaa1248, pubchem19048, z-o-benzyl-l-threonine
PubChem CID 7018810
IUPAC Name (2S,3R)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6532603 View Documents Alfa Aesar™
H6532603
1g Each for $34.55
 
AAH6532606 View Documents Alfa Aesar™
H6532606
5g Each for $149.45
 
AAH6532614 View Documents Alfa Aesar™
H6532614
25g Each for $619.93
 
Specifications

Chemical Identifiers

69863-36-1
343.379
FVKJXAYBJIAXAU-PBHICJAKSA-N
7018810
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OCC2=CC=CC=C2
C19H21NO5
MFCD00077042
z-thr bzl-oh, n-z-o-benzyl-l-threonine, n-cbz-o-benzyl-l-threonine, cbz-thr bzl-oh, o-benzyl-n-benzyloxycarbonyl-l-threonine, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, ambotzzaa1248, pubchem19048, z-o-benzyl-l-threonine
(2S,3R)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

69863-36-1
MFCD00077042
z-thr bzl-oh, n-z-o-benzyl-l-threonine, n-cbz-o-benzyl-l-threonine, cbz-thr bzl-oh, o-benzyl-n-benzyloxycarbonyl-l-threonine, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, 2s,3r-3-benzyloxy-2-benzyloxy carbonyl amino butanoic acid, ambotzzaa1248, pubchem19048, z-o-benzyl-l-threonine
CC(C(C(=O)O)NC(=O)OCC1=CC=CC=C1)OCC2=CC=CC=C2
343.379
343.38
95%
1g
C19H21NO5
5988744
FVKJXAYBJIAXAU-PBHICJAKSA-N
(2S,3R)-3-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
7018810
Solid
77°C to 80°C
O-Benzyl-N-benzyloxycarbonyl-L-threonine