missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%

$31.76 - $124.77

Chemical Identifiers

CAS 34805-17-9
Molecular Formula C17H20ClNO3
Molecular Weight (g/mol) 321.801
MDL Number MFCD00080888
InChI Key IQKXGACPIPNLEL-NTISSMGPSA-N
Synonym h-tyr bzl-ome.hcl, h-tyr bzl-ome hcl, o-benzyl-l-tyrosine methyl ester hydrochloride, c17h19no3.hcl, l-tyrosine, o-phenylmethyl-, methyl ester, hydrochloride, h-tyr bzl-omehcl, h-tyr bzl-ome??hcl, h-tyr bzl-ome·hcl, h-tyr bzl-ome hydrochloride, o-benzyl tyrosine methyl ester hydrochloride
PubChem CID 12616037
IUPAC Name methyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
SMILES COC(=O)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6390803 View Documents Alfa Aesar™
H6390803
1g Each for $31.76
 
AAH6390806 View Documents Alfa Aesar™
H6390806
5g Each for $124.77
 
Specifications

Chemical Identifiers

34805-17-9
321.801
IQKXGACPIPNLEL-NTISSMGPSA-N
12616037
COC(=O)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)N.Cl
C17H20ClNO3
MFCD00080888
h-tyr bzl-ome.hcl, h-tyr bzl-ome hcl, o-benzyl-l-tyrosine methyl ester hydrochloride, c17h19no3.hcl, l-tyrosine, o-phenylmethyl-, methyl ester, hydrochloride, h-tyr bzl-omehcl, h-tyr bzl-ome??hcl, h-tyr bzl-ome·hcl, h-tyr bzl-ome hydrochloride, o-benzyl tyrosine methyl ester hydrochloride
methyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride

Specifications

34805-17-9
MFCD00080888
IQKXGACPIPNLEL-NTISSMGPSA-N
methyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
12616037
Solid
1g
C17H20ClNO3
h-tyr bzl-ome.hcl, h-tyr bzl-ome hcl, o-benzyl-l-tyrosine methyl ester hydrochloride, c17h19no3.hcl, l-tyrosine, o-phenylmethyl-, methyl ester, hydrochloride, h-tyr bzl-omehcl, h-tyr bzl-ome??hcl, h-tyr bzl-ome·hcl, h-tyr bzl-ome hydrochloride, o-benzyl tyrosine methyl ester hydrochloride
COC(=O)C(CC1=CC=C(C=C1)OCC2=CC=CC=C2)N.Cl
321.801
321.8
98%
O-Benzyl-L-tyrosine methyl ester hydrochloride
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C