missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Benzyl-L-tyrosine benzyl ester hydrochloride, 95%, Thermo Scientific™
$50.44 - $185.88
Chemical Identifiers
CAS | 52142-01-5 |
---|---|
Molecular Formula | C23H24ClNO3 |
Molecular Weight (g/mol) | 397.90 |
MDL Number | MFCD00153465 |
InChI Key | DYLKTBYVTWFYGQ-DKVUPULANA-N |
Synonym | h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride |
PubChem CID | 45073233 |
IUPAC Name | benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride |
SMILES | Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6315603
|
Thermo Scientific Chemicals
H6315603 |
1 g |
Each for $50.44
|
|
|||||
AAH6315606
|
Thermo Scientific Chemicals
H6315606 |
5 g |
Each for $185.88
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
52142-01-5 | |
397.90 | |
DYLKTBYVTWFYGQ-DKVUPULANA-N | |
45073233 | |
Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1 |
C23H24ClNO3 | |
MFCD00153465 | |
h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride | |
benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride |
Specifications
52142-01-5 | |
MFCD00153465 | |
DYLKTBYVTWFYGQ-DKVUPULANA-N | |
benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride | |
45073233 | |
95% | |
Solid |
C23H24ClNO3 | |
h-tyr bzl-obzl.hcl, h-tyr bzl-obzl hcl, o-benzyl-l-tyrosine benzyl ester hydrochloride, l-tyrosine, o-phenylmethyl-, phenylmethyl ester, hydrochloride, httyr bzl-obzl hcl, h-tyr bzl-obzlcl, h-tyr bzl-obzl??hcl, h-tyr bzl-obzl hydrochloride, benzyl o-benzyl-l-tyrosinate-hydrogen chloride 1/1, benzyl 2s-2-amino-3-4-benzyloxyphenyl propanoate hydrochloride | |
Cl.N[C@@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(=O)OCC1=CC=CC=C1 | |
397.90 | |
397.89 | |
1 g | |
O-Benzyl-L-tyrosine benzyl ester hydrochloride |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold