missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-L-tyrosine, 98%

Manufacturer:  Alfa Aesar™ H6367522

 View more versions of this product

Catalog No. AAH6367522


 

Specifications

Specifications

O-Benzyl-L-tyrosine
98%
MFCD00002605
H-Tyr(Bzl)-OH; 4-Benzyloxy-L-phenylalanine
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)N
271.316
271.31
White
100g
16652-64-5
C16H17NO3
2659642
KAFHLONDOVSENM-HNNXBMFYSA-N
(2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoic acid
725701
Powder
259°C (decomposition)

Chemical Identifiers

16652-64-5
271.316
KAFHLONDOVSENM-HNNXBMFYSA-N
725701
C16H17NO3
MFCD00002605
H-Tyr(Bzl)-OH; 4-Benzyloxy-L-phenylalanine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents