Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-L-threonine hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material O-Benzyl-L-threonine hydrochloride
CAS 60856-51-1
Formula Weight 245.7
MDL Number MFCD00037768
Synonym H-Thr(Bzl)-OH.HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6598003

  SDS
alfa aesar™
H6598003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6598006

  SDS
alfa aesar™
H6598006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.7
Chemical Name or Material O-Benzyl-L-threonine hydrochloride
MDL Number MFCD00037768
CAS 60856-51-1
Physical Form Solid
Storage Note 1 2° to 8°C
Quantity 1g
Assay 95%
Synonym H-Thr(Bzl)-OH.HCl
Safety and Handling

2° to 8°C