missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Benzyl-L-serine Methyl Ester Hydrochloride 98.0+%, TCI America™
$149.32 - $462.86
Chemical Identifiers
CAS | 19525-87-2 |
---|---|
Molecular Formula | C11H16ClNO3 |
Molecular Weight (g/mol) | 245.70 |
MDL Number | MFCD00191312 |
InChI Key | UDHSAWHYRXHQJP-UHFFFAOYNA-N |
Synonym | Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl |
PubChem CID | 44630138 |
IUPAC Name | methyl 2-amino-3-(benzyloxy)propanoate hydrochloride |
SMILES | Cl.COC(=O)C(N)COCC1=CC=CC=C1 |
Chemical Identifiers
19525-87-2 | |
245.70 | |
UDHSAWHYRXHQJP-UHFFFAOYNA-N | |
44630138 | |
Cl.COC(=O)C(N)COCC1=CC=CC=C1 |
C11H16ClNO3 | |
MFCD00191312 | |
Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl | |
methyl 2-amino-3-(benzyloxy)propanoate hydrochloride |
Specifications
19525-87-2 | |
C11H16ClNO3 | |
1 g | |
UDHSAWHYRXHQJP-UHFFFAOYNA-N | |
methyl 2-amino-3-(benzyloxy)propanoate hydrochloride | |
44630138 | |
≥98.0% (T) | |
O-Benzyl-L-serine Methyl Ester Hydrochloride |
White | |
MFCD00191312 | |
Methyl O-Benzyl-L-serinate Hydrochloride, H-Ser(Bzl)-OMe.HCl | |
Cl.COC(=O)C(N)COCC1=CC=CC=C1 | |
245.70 | |
245.70 | |
Crystalline Powder |
Safety and Handling
TSCA : No