missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Benzyl-L-serine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2239314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
213°C (decomposition) | |
4726-96-9 | |
MFCD00065937 | |
o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine | |
C1=CC=C(C=C1)COCC(C(=O)O)N | |
195.218 | |
195.22 | |
25 g |
O-Benzyl-L-serine | |
C10H13NO3 | |
2114846 | |
IDGQXGPQOGUGIX-VIFPVBQESA-N | |
(2S)-2-amino-3-phenylmethoxypropanoic acid | |
78457 | |
99% | |
+7° (c=1 in 1N HCl) |
Chemical Identifiers
4726-96-9 | |
195.218 | |
IDGQXGPQOGUGIX-VIFPVBQESA-N | |
78457 | |
C1=CC=C(C=C1)COCC(C(=O)O)N |
C10H13NO3 | |
MFCD00065937 | |
o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine | |
(2S)-2-amino-3-phenylmethoxypropanoic acid |
Safety and Handling
EINECSNumber : 225-220-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only