Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-DL-serine

$38.95 - $113.59

Chemical Identifiers

Chemical Name or Material O-Benzyl-DL-serine
CAS 5445-44-3
Molecular Formula C10H13NO3
Formula Weight 195.22
MDL Number MFCD00021724
Beilstein 3204905
Synonym (+/-)-2-Amino-3-(benzyloxy)propionic acid; H-DL-Ser(Bzl)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1949103

 
alfa aesar™
L1949103
1g Each for $38.95

AAL1949106

 
alfa aesar™
L1949106
5g Each for $113.59
Description & Specifications

Specifications

Formula Weight 195.22
Chemical Name or Material O-Benzyl-DL-serine
Molecular Formula C10H13NO3
MDL Number MFCD00021724
CAS 5445-44-3
Beilstein 3204905
Synonym (+/-)-2-Amino-3-(benzyloxy)propionic acid; H-DL-Ser(Bzl)-OH
Melting Point ≈225°C
Quantity 1g

Applications
O-Benzyl-DL-serine is used as catalytic agent, petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Store away from oxidizing agents.