missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-Benzyl-DL-serine, TCI America™

$47.36 - $209.31

Chemical Identifiers

CAS 5445-44-3
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00021724
InChI Key IDGQXGPQOGUGIX-UHFFFAOYSA-N
Synonym o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
PubChem CID 4573
IUPAC Name 2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B08601G View Documents TCI America
B08601G
1g Each for $47.36
 
B086010G View Documents TCI America
B086010G
10g Each for $209.31
 
Specifications

Chemical Identifiers

5445-44-3
195.218
IDGQXGPQOGUGIX-UHFFFAOYSA-N
4573
C1=CC=C(C=C1)COCC(C(=O)O)N
C10H13NO3
MFCD00021724
o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
2-amino-3-phenylmethoxypropanoic acid

Specifications

White-Yellow
5445-44-3
MFCD00021724
IDGQXGPQOGUGIX-UHFFFAOYSA-N
2-amino-3-phenylmethoxypropanoic acid
4573
Crystal-Powder at 20°C
O-Benzyl-DL-serine
1g
C10H13NO3
o-benzyl-dl-serine, 2-amino-3-benzyloxy propanoic acid, o-benzylserine, h-dl-ser bzl-oh, 3-benzyloxy-dl-alanine, benzylserine, 2-amino-3-phenylmethoxy propanoic acid, o-benzyl-serine, acmc-1artk, spectrum2_001351
C1=CC=C(C=C1)COCC(C(=O)O)N
195.218
195.22
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No