Please login to your online account to display your discounted pricing

Alfa Aesar™ O-Benzyl-D-tyrosine, 95%

Chemical Identifiers

Chemical Name or Material O-Benzyl-D-tyrosine
CAS 65733-15-5
Formula Weight 271.32
MDL Number MFCD00077104
Beilstein 5819231
Synonym H-D-Tyr(Bzl)-OH; (R)-2-Amino-3-[4-(benzyloxy)phenyl]propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6500203

 
alfa aesar™
H6500203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6500206

 
alfa aesar™
H6500206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6500214

 
alfa aesar™
H6500214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 271.32
Chemical Name or Material O-Benzyl-D-tyrosine
CAS 65733-15-5
Physical Form Solid
MDL Number MFCD00077104
Beilstein 5819231
Assay 95%
Synonym H-D-Tyr(Bzl)-OH; (R)-2-Amino-3-[4-(benzyloxy)phenyl]propionic acid
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C