missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
O-Benzoyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™
$87.92 - $350.05
Chemical Identifiers
CAS | 27130-46-7 |
---|---|
Molecular Formula | C8H10ClNO2 |
Molecular Weight (g/mol) | 187.623 |
InChI Key | FTCDQNQIGUCFSA-UHFFFAOYSA-N |
PubChem CID | 11615307 |
IUPAC Name | methylamino benzoate;hydrochloride |
SMILES | CNOC(=O)C1=CC=CC=C1.Cl |
Chemical Identifiers
27130-46-7 | |
187.623 | |
11615307 | |
CNOC(=O)C1=CC=CC=C1.Cl |
C8H10ClNO2 | |
FTCDQNQIGUCFSA-UHFFFAOYSA-N | |
methylamino benzoate;hydrochloride |
Specifications
27130-46-7 | |
White-Yellow | |
1 g | |
CNOC(=O)C1=CC=CC=C1.Cl | |
187.623 | |
187.62 | |
Crystalline Powder |
136°C | |
C8H10ClNO2 | |
FTCDQNQIGUCFSA-UHFFFAOYSA-N | |
methylamino benzoate;hydrochloride | |
11615307 | |
≥98.0% (HPLC,N) | |
O-Benzoyl-N-methylhydroxylamine Hydrochloride |
Safety and Handling
TSCA : No