missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Acros Organics

Chemical Identifiers

Assay Percent Range 98%
Molecular Formula C11H16N5O·PF6
Formula Weight 379.25
Synonym O-Benzotriazol-1-yl-tetramethyluronium hexafluorophosphate
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC299430010 Thermo Fisher Scientific
AC299430010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC299430050 Thermo Fisher Scientific
AC299430050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC299430250 Thermo Fisher Scientific
AC299430250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Coupling reagent
Description & Specifications

Chemical Identifiers

98%
379.25
O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
C11H16N5O·PF6
O-Benzotriazol-1-yl-tetramethyluronium hexafluorophosphate

Specifications

379.25
94790-37-1
98%
Glass bottle
1g
C11H16N5O·PF6
O-Benzotriazol-1-yl-tetramethyluronium hexafluorophosphate
O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate