Please login to your online account to display your discounted pricing

O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%, Acros Organics


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC299430250

 View more versions of this product

Catalog No. AC299430250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 379.25
Quantity 25g
CAS 94790-37-1
Synonym O-Benzotriazol-1-yl-tetramethyluronium hexafluorophosphate
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H16N5O·PF6

Specifications

Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
Assay Percent Range 98%
Molecular Formula C11H16N5O·PF6
Formula Weight 379.25
Synonym O-Benzotriazol-1-yl-tetramethyluronium hexafluorophosphate