missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium Hexafluorophosphate 98.0+%, TCI America™

$56.41 - $242.21

Chemical Identifiers

CAS 105379-24-6
Molecular Formula C15H20F6N5OP
Molecular Weight (g/mol) 431.323
MDL Number MFCD00191770
InChI Key XKTRAGMCMJYRRN-UHFFFAOYSA-N
Synonym hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
PubChem CID 13685355
IUPAC Name 1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
SMILES C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B48051G
View Documents Promotion Details
TCI America
B48051G
1 g
Each for $56.41
 
B48055G
View Documents Promotion Details
TCI America
B48055G
5 g
Each for $242.21
 
Description

Description

Specifications

Chemical Identifiers

105379-24-6
431.323
XKTRAGMCMJYRRN-UHFFFAOYSA-N
13685355
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
C15H20F6N5OP
MFCD00191770
hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate

Specifications

105379-24-6
White
MFCD00191770
hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
431.323
431.32
Crystalline Powder
171°C
C15H20F6N5OP
1 g
XKTRAGMCMJYRRN-UHFFFAOYSA-N
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
13685355
≥98.0% (HPLC,N)
O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(tetramethylene)uronium Hexafluorophosphate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'