missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440250050

 View more versions of this product

Catalog No. AC440250050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Authentic
176°C
105379-24-6
100.0
C15H20N5O·F6P
XKTRAGMCMJYRRN-UHFFFAOYSA-N
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
13685355
Powder
40.73 to 42.81% C, 15.83 to 16.64% N
White
5g
97.5
98%
1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, hbpyu, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
431.32
431.32
98%

Chemical Identifiers

105379-24-6
431.32
1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, hbpyu, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
C15H20N5O·F6P
XKTRAGMCMJYRRN-UHFFFAOYSA-N
13685355
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents