missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(pentamethylene)uronium Hexafluorophosphate, TCI America™

$91.67 - $311.67

Chemical Identifiers

CAS 206752-41-2
Molecular Formula C17H24F6N5OP
Molecular Weight (g/mol) 459.377
MDL Number MFCD00191769
InChI Key YNOBMGHLCWIWCL-UHFFFAOYSA-N
Synonym (Benzotriazol-1-yloxy)dipiperidinocarbenium Hexafluorophosphate, HBPipU
PubChem CID 10863473
IUPAC Name 1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
SMILES C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B38161G View Documents TCI America
B38161G
1g Each for $91.67
 
B38165G View Documents TCI America
B38165G
5g Each for $311.67
 
Specifications

Chemical Identifiers

206752-41-2
459.377
YNOBMGHLCWIWCL-UHFFFAOYSA-N
10863473
C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
C17H24F6N5OP
MFCD00191769
(Benzotriazol-1-yloxy)dipiperidinocarbenium Hexafluorophosphate, HBPipU
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate

Specifications

White
1g
C17H24F6N5OP
(Benzotriazol-1-yloxy)dipiperidinocarbenium Hexafluorophosphate, HBPipU
C1CCN(CC1)C(=[N+]2CCCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
459.377
459.38
≥98.0% (HPLC)
199°C
206752-41-2
MFCD00191769
YNOBMGHLCWIWCL-UHFFFAOYSA-N
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
10863473
Crystal-Powder at 20°C
O-(Benzotriazol-1-yl)-N,N,N′,N′-bis(pentamethylene)uronium Hexafluorophosphate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator