Learn More
o-Benzoic sulfimide sodium salt hydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1553030
Description
O-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsO-benzoic sulfimide sodium salt is used as an artificial sweetener in drinks, candies, cookies, medicines and toothpaste. It is used in the food industry and for sweetening diabetes diet. It is also used as a synthetic sweet ignorance agent. It finds application in a variety of beverages, foods, cosmetics and pharmaceuticals to provide sweetness without adding calories. It is involved as a nickel electroplating brightener, chemical intermediate, animal feed sweetener and anaerobic adhesive accelerator.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
o-Benzoic sulfimide sodium salt hydrate | |
>300°C | |
MFCD00149605 | |
3599229 | |
saccharin sodium salt hydrate | |
OAZGZMKGRPRHJX-UHFFFAOYSA-M | |
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate | |
131673955 | |
99% |
82385-42-0 | |
C7H4NNaO3S | |
250 g | |
14,8312 | |
Soluble in water. | |
[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 | |
205.16 | |
205.16 (Anhydrous) |
Chemical Identifiers
82385-42-0 | |
205.16 | |
OAZGZMKGRPRHJX-UHFFFAOYSA-M | |
131673955 | |
[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12 |
C7H4NNaO3S | |
MFCD00149605 | |
saccharin sodium salt hydrate | |
1,1-dioxo-1,2-benzothiazol-3-one;molecular hydrogen;sodium;hydrate |
Safety and Handling
EINECSNumber : 204-886-1
RTECSNumber : DE4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only